Learn More
Quality Biological Inc Serological Pipet, 2 mL, Polystryrene, Individually wrapped, Sterile, packed 100/bag, 600/CS,
SDP
Catalog No. 507517908
Encompass_Preferred
Serological Pipet, 2 mL, Polystryrene, Individually wrapped, Sterile, Packed 100/bag, 600/CS
• Manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification
Sterile: Yes
Maximum Volume: 2 mL
Graduated: Yes
Graduation Increments (Subdivisions): 1/10 mL
Material: Polystyrene
Catalog No. 50-751-7908 Supplier Quality Biological Inc Supplier No. P886229202B
Serological Pipet, 2 mL, Polystryrene, Individually wrapped, Sterile, Packed 100/bag, 600/CS
• Manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification
Sterile: Yes
Maximum Volume: 2 mL
Graduated: Yes
Graduation Increments (Subdivisions): 1/10 mL
Material: Polystyrene