Learn More
Quality Biological Inc Pipet 5 0mL, Sterile, Bulk Packed, 25 per bag, 200/CS
SDP
Catalog No. 507517800
Encompass_Preferred
Pipet 50 mL, Sterile, Bulk Packed, Packaged 25 bulk packed/Bag, 200/CS
• Convenient bulk packaging option
• All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification
Catalog No. 50-751-7800 Supplier Quality Biological Inc Supplier No. P886229250
Pipet 50 mL, Sterile, Bulk Packed, Packaged 25 bulk packed/Bag, 200/CS
• Convenient bulk packaging option
• All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification