Learn More
Quality Biological Inc 50 mL Pipet, Individually Wrapped, Sterile, Packed in Bags, 30/Bag, 90/CS
SDP
Catalog No. 507517873
Encompass_Preferred
50 mL Pipet, Individually Wrapped, Sterile, Packed in Bags
Packaged 30 Individually Wrapped/Bag, 90/CS
• All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification
Catalog No. 50-751-7873 Supplier Quality Biological Inc Supplier No. P886229230B
50 mL Pipet, Individually Wrapped, Sterile, Packed in Bags
Packaged 30 Individually Wrapped/Bag, 90/CS
• All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification