Learn More
Quality Biological Inc Serological Pipet, 5 mL, Polystyrene, Individually wrapped, Sterile, packed 50/bag, 200/CS
SDP
Catalog No. 507517872
Encompass_Preferred
Serological Pipet, 5 mL, Polystyrene, Individually wrapped, Sterile, Packed 50/bag, 200/CS
• Manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification
Sterile: Yes
Maximum Volume: 5 mL
Graduated: Yes
Graduation Increments (Subdivisions): 1/10 mL
Material: Polystyrene
Catalog No. 50-751-7872 Supplier Quality Biological Inc Supplier No. P886229205B
Serological Pipet, 5 mL, Polystyrene, Individually wrapped, Sterile, Packed 50/bag, 200/CS
• Manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification
Sterile: Yes
Maximum Volume: 5 mL
Graduated: Yes
Graduation Increments (Subdivisions): 1/10 mL
Material: Polystyrene