Learn More
Quality Biological Inc Plasteur Polystyrene Pasteur Pipet, 6 Inch Length, Sterile, Individually Wrapped, 50/Bag, 200/CS
SDP
Catalog No. 50147427
Encompass_Preferred
Plasteur Polystyrene Pasteur Pipet, 5.75 Inch Length, Sterile, Individually Wrapped
Packaged 50 Individually Wrapped/Bag, 200/CS
• The Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plastic
• Fine tip is ideal for precision aspirating
• No more broken glass: more durable than standard glass pipets
• No damage during shipping and handling as compared to traditional glass pipets
• Available sterile for convenience
Catalog No. 50-147-427 Supplier Quality Biological Inc Supplier No. P886229270
Plasteur Polystyrene Pasteur Pipet, 5.75 Inch Length, Sterile, Individually Wrapped
Packaged 50 Individually Wrapped/Bag, 200/CS
• The Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plastic
• Fine tip is ideal for precision aspirating
• No more broken glass: more durable than standard glass pipets
• No damage during shipping and handling as compared to traditional glass pipets
• Available sterile for convenience