Learn More
Quality Biological Inc Plasteur Polystyrene Pasteur Pipet, 6 Inch Length, Sterile, Bulk Packed, 25/Bag, 500/CS
SDP
Catalog No. 50147428
Encompass_Preferred
Plasteur Polystyrene Pasteur Pipet, 5.75 Inch Length, Sterile, Bulk Packed
Packaged 25 Bulk Packed/Bag, 500/CS
• The Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plastic
• Fine tip is ideal for precision aspirating
• No more broken glass: more durable than standard glass pipets
• No damage during shipping and handling as compared to traditional glass pipets
• Available sterile for convenience
Catalog No. 50-147-428 Supplier Quality Biological Inc Supplier No. P886229275
Plasteur Polystyrene Pasteur Pipet, 5.75 Inch Length, Sterile, Bulk Packed
Packaged 25 Bulk Packed/Bag, 500/CS
• The Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plastic
• Fine tip is ideal for precision aspirating
• No more broken glass: more durable than standard glass pipets
• No damage during shipping and handling as compared to traditional glass pipets
• Available sterile for convenience