Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Pipet Adapter

Catalog No. FB14955203 Shop All Fisherbrand Products
Change view
Click to view available options
Pipet Adapter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
FB14955203 Pipet Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FB14955203
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement Pipet Adapter for use with Fisherbrand Pipet Controller

Silicone pipet adaptor with safety valve

Specifications

Description Replacement parts only
Color Gray
For Use With (Equipment) Fisherbrand Pipet Controller (FB14955202)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.