Learn More
Sigma Aldrich Fine Chemicals Biosciences Pipet AidR automatic pipet1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P18251EA
Encompass
Safe convenient instruments for converting pipettes into hand-operated dispensers. Especially useful in confined areas such as laminar flow hoods. Can be used with glass or plastic pipettes ranging in size from 1 to 100 mL.
Catalog No. 50-290-9047