Learn More
Sigma Aldrich Fine Chemicals Biosciences Pipet AidR automatic pipet1EA
Catalog No. 502904842
Encompass
Safe convenient instruments for converting pipettes into hand-operated dispensers. Especially useful in confined areas such as laminar flow hoods. Can be used with glass or plastic pipettes ranging in size from 1 to 100 mL.
Catalog No. 50-290-4842 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P61751EA
Safe convenient instruments for converting pipettes into hand-operated dispensers. Especially useful in confined areas such as laminar flow hoods. Can be used with glass or plastic pipettes ranging in size from 1 to 100 mL.