Learn More
CELLTREAT Scientific Products 50mL Aspirating Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile
SDP
Catalog No. NC1964623
Encompass_Preferred
Ideal for aspiration of media and sample waste removal without needing to measure the volume. Aspirating Pipets are supplied without graduations or plugs. Also available in 10, 25 and 50mL Individually Wrapped sizes. 2mL Aspirating Pipet available in plastic/plastic wrap (Part No. 229262A). Peel-to-open plastic/plastic wrappers are the ideal option for eliminating particulate; proven benefit over traditional paper/plastic wrapper.
Catalog No. NC1964623 Supplier CELLTREAT Scientific Products Supplier No. 229249
Ideal for aspiration of media and sample waste removal without needing to measure the volume. Aspirating Pipets are supplied without graduations or plugs. Also available in 10, 25 and 50mL Individually Wrapped sizes. 2mL Aspirating Pipet available in plastic/plastic wrap (Part No. 229262A). Peel-to-open plastic/plastic wrappers are the ideal option for eliminating particulate; proven benefit over traditional paper/plastic wrapper.