Learn More
Integra Biosciences Corp PIPETBOY acu 2 pipet aid, Green
SDP
Catalog No. NC0165100
Encompass
Pipet Aid complete with battery Lithium Polymer, mains adapter, spare sterile filter 0.45 um and wall mount. Color: green. The second generation of PIPETBOY acu pipet aid is equipped with the latest technology providing an increased pipetting speed and a significant weight reduction of the instrument. These two features help you to cut your work time and offer you an improved ergonomic pipetting experience.
Catalog No. NC0165100 Supplier Integra Biosciences Corp Supplier No. 155015
Pipet Aid complete with battery Lithium Polymer, mains adapter, spare sterile filter 0.45 um and wall mount. Color: green. The second generation of PIPETBOY acu pipet aid is equipped with the latest technology providing an increased pipetting speed and a significant weight reduction of the instrument. These two features help you to cut your work time and offer you an improved ergonomic pipetting experience.