Learn More
Integra Biosciences Corp PIPETBOY acu 2 Pipet Aid, Purple
SDP
Catalog No. NC0085686
Encompass
The second generation of PIPETBOY acu pipet aid is equipped with the latest technology providing an increased pipetting speed and a significant weight reduction of the instrument. These two features help you to cut your work time and offer you an improved ergonomic pipetting experience. Besides these changes the main benefits of PIPETBOY acu were preserved: the unmatched pipetting precision, the colorful look and the weight balance. Maximum pipetting speed 13.5 ml/s:(with a 50 ml pipette). Gravity dispense Slight pressure on dispense button. Pipette compatibility Glass and plastic pipettes with volume from 1 to 100 ml.
This item is not returnable. View return policy
Catalog No. NC0085686 Supplier Integra Biosciences Corp Supplier No. 155019
The second generation of PIPETBOY acu pipet aid is equipped with the latest technology providing an increased pipetting speed and a significant weight reduction of the instrument. These two features help you to cut your work time and offer you an improved ergonomic pipetting experience. Besides these changes the main benefits of PIPETBOY acu were preserved: the unmatched pipetting precision, the colorful look and the weight balance. Maximum pipetting speed 13.5 ml/s:(with a 50 ml pipette). Gravity dispense Slight pressure on dispense button. Pipette compatibility Glass and plastic pipettes with volume from 1 to 100 ml.