Learn More
Chemglass Life Sciences Pipet Bulb, Latex, 2mL, Natural Color
SDP
Catalog No. 501897606
Encompass
Capacity: 2mL (1/15oz) Bore Size: 6mm (1/4in) Length: 45mm (1-3/4in) Diameter: 18mm (5/8in)
Catalog No. 50-189-7606 Supplier Chemglass Life Sciences Supplier No. CGE3488C10
Capacity: 2mL (1/15oz) Bore Size: 6mm (1/4in) Length: 45mm (1-3/4in) Diameter: 18mm (5/8in)