Learn More
CELLTREAT Scientific Products Electronic Pipet Controller
SDP
Catalog No. 50550475
Encompass_Preferred
CELLTREAT Scientific Products Pipet Controller, 120V. Lightweight (6.3oz/170g) and designed to fit comfortably in either hand. Long-life rechargeable lithium battery provides up to 8 hours of cordless use on a 2 hour charge. Adjustable dial for speed setting. Low pressure fingertip control buttons. Easy fit adapter accepts pipet sizes from 1 to 100mL. Every case is printed with the lot number for quality traceability. Individually packed 1/case.
Catalog No. 50-550-475 Supplier CELLTREAT Scientific Products Supplier No. 230200
CELLTREAT Scientific Products Pipet Controller, 120V. Lightweight (6.3oz/170g) and designed to fit comfortably in either hand. Long-life rechargeable lithium battery provides up to 8 hours of cordless use on a 2 hour charge. Adjustable dial for speed setting. Low pressure fingertip control buttons. Easy fit adapter accepts pipet sizes from 1 to 100mL. Every case is printed with the lot number for quality traceability. Individually packed 1/case.