Learn More
CELLTREAT Scientific Products Pipet Controller, Manual, 1/CS
SDP
Catalog No. 501535085
Encompass_Preferred
CELLTREAT Scientific Products Manual Pipet Controller. Lightweight manual pipet controller does not require batteries. Squeeze bulb creates pressure to draw fluid up into pipets; up to 50mL per squeeze. Ergonomic thumb lever controls speed of aspirating and dispensing. Press button blows out remaining drops in pipet. Compatible with 1-100mL serological pipets.
Catalog No. 50-153-5085 Supplier CELLTREAT Scientific Products Supplier No. 230210
Add to Cart
Add to Cart
CELLTREAT Scientific Products Manual Pipet Controller. Lightweight manual pipet controller does not require batteries. Squeeze bulb creates pressure to draw fluid up into pipets; up to 50mL per squeeze. Ergonomic thumb lever controls speed of aspirating and dispensing. Press button blows out remaining drops in pipet. Compatible with 1-100mL serological pipets.