Learn More
Bellco Glass, Inc. Pipet Controller Manual
SDP
Catalog No. 502330629
Encompass_Preferred
CELLTREAT Lightweight manual pipet controller does not require batteries. Squeeze bulb creates pressure to draw fluid up into pipets up to 50mL per squeeze. Ergonomic thumb lever controls speed of aspirating and dispensing.Specifications Press button blows out remaining drops in pipet Easy fit adapter accepts pipet sizes from 1 to 100mL
Catalog No. 50-233-0629 Supplier Bellco Glass, Inc. Supplier No. 230210000
CELLTREAT Lightweight manual pipet controller does not require batteries. Squeeze bulb creates pressure to draw fluid up into pipets up to 50mL per squeeze. Ergonomic thumb lever controls speed of aspirating and dispensing.Specifications Press button blows out remaining drops in pipet Easy fit adapter accepts pipet sizes from 1 to 100mL