Learn More
Hach Company Pipet Filler-3-Valve
Supplier:  Hach Company 1218900
Encompass
Easy 1-2-3 operation Squeeze the 1 valve to discharge air in top bulb Squeeze valve 2 to draw up sample Squeeze valve 3 to dispense Fits all standard-sized pipets Durable silicone for long life Meets OSHA requirements Accommodates pipettes up to 50mL and has a 1 4 in diameter opening
Catalog No. 50-002-57101