Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pipet Filler
SCI_ED

Catalog No. S32245 Shop All Tip Industries Inc. Products
Change view
Click to view available options
Color:
Black
Red
Material:
Rubber
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
S32245 Red Rubber
S32244 Black Rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S32245 Supplier Tip Industries, Inc. Supplier No. 5030RED
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fits pipets up to 50mL.

  • Precise to 0.01mL
  • Chemical-resistant rubber
  • Expel air by pressing valve A on top of the bulb
  • Draw liquid up by pressing S valve on stem
  • To release, press valve E on side of the stem

Specifications

Autoclavable Not Autoclavable
Color Red
Material Rubber
Type Pipette Filler
For Use With (Equipment) Pipet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.