Learn More
Pipette Supplies, Inc. PIPET-LITE O-RING 200UL 5PK
SDP
Catalog No. 501317567
Encompass
Piston O-ring for the following Rainin 200μL Pipettes:Pipet-Lite (L, SL)Pipet-Lite XLS (XLS)Pipet-Plus (R, RL)EDP1 (E1, SE1)EDP3, EDP3 Plus (E3, SE3)E-Man Hybrid (H, SH)E4 XLS (E4, SE4)Alternative to Rainin 6200-155 / 17003378
Catalog No. 50-131-7567 Supplier Pipette Supplies, Inc. Supplier No. 20200/5
Piston O-ring for the following Rainin 200μL Pipettes:Pipet-Lite (L, SL)Pipet-Lite XLS (XLS)Pipet-Plus (R, RL)EDP1 (E1, SE1)EDP3, EDP3 Plus (E3, SE3)E-Man Hybrid (H, SH)E4 XLS (E4, SE4)Alternative to Rainin 6200-155 / 17003378