Learn More
Pipette Supplies, Inc. PIPET-LITE O-RING 2UL 5PK
SDP
Catalog No. 501317563
Encompass
Piston o-ring for:Pipet-Lite (L, SL)Pipet-Lite XLS (XLS)Pipet-Plus (R, RL)EDP1 (E1, SE1)EDP3, EDP3 Plus (E3, SE3)E-Man Hybrid (H, SH)Alternative to Rainin 6200-132 / 17003372
Catalog No. 50-131-7563 Supplier Pipette Supplies, Inc. Supplier No. 20002/5
Piston o-ring for:Pipet-Lite (L, SL)Pipet-Lite XLS (XLS)Pipet-Plus (R, RL)EDP1 (E1, SE1)EDP3, EDP3 Plus (E3, SE3)E-Man Hybrid (H, SH)Alternative to Rainin 6200-132 / 17003372