Learn More
Pipette Supplies, Inc. PIPET-LITE, PIPET-PLUS CALIBRA
SDP
Catalog No. NC0366305
Encompass
Calibration Tool designed for Rainin pipettes: Pipet-Lite, Pipet-Lite XLS and the Pipet-Plus. Top Seller. Instructions included. Two piece set. 304 steel. One steel cylinder goes inside other cylinder. Together, these cylinders slide down the plunger rod for a calibration adjustment. Use both pieces when adjusting the Pipet-Lite, Pipet-Lite XLS or Pipet-Plus. Use only the inner cylinder to adjust the pre-Jan 1986 style (6 hex screws) Pipetman.
Catalog No. NC0366305 Supplier Pipette Supplies, Inc. Supplier No. RA CT2
Calibration Tool designed for Rainin pipettes: Pipet-Lite, Pipet-Lite XLS and the Pipet-Plus. Top Seller. Instructions included. Two piece set. 304 steel. One steel cylinder goes inside other cylinder. Together, these cylinders slide down the plunger rod for a calibration adjustment. Use both pieces when adjusting the Pipet-Lite, Pipet-Lite XLS or Pipet-Plus. Use only the inner cylinder to adjust the pre-Jan 1986 style (6 hex screws) Pipetman.