Learn More
Pipette Supplies, Inc. PIPET-LITE SEAL 200UL, 5PK
SDP
Catalog No. 501317615
Encompass
Piston seal for the following Rainin 200μL Pipettes:Pipet-Lite (L, SL)Pipet-Lite XLS (XLS)Pipet-Plus (R, RL)EDP1 (E1, SE1)EDP3, EDP3 Plus (E3, SE3)E-Man Hybrid (H, SH)E4 XLS (E4, SE4)Alternative to Rainin 6200-154 / 17004642
Catalog No. 50-131-7615 Supplier Pipette Supplies, Inc. Supplier No. 80200/5
Piston seal for the following Rainin 200μL Pipettes:Pipet-Lite (L, SL)Pipet-Lite XLS (XLS)Pipet-Plus (R, RL)EDP1 (E1, SE1)EDP3, EDP3 Plus (E3, SE3)E-Man Hybrid (H, SH)E4 XLS (E4, SE4)Alternative to Rainin 6200-154 / 17004642