Learn More
Pipette Supplies, Inc. Rainin Pipet-Lite XLS+ Grease, 45 grams (Pipette Supplies)
SDP
Catalog No. NC1844271
Encompass
Used with E4 XLS+ & Pipet-LiteXLS+ pipettes up to 2000ul, and all previous pipettes being converted to the XLS+ sealing system. Identical silicone lubricant sold by Rainin.
Catalog No. NC1844271 Supplier Pipette Supplies, Inc. Supplier No. 2XLSGR55
Used with E4 XLS+ & Pipet-LiteXLS+ pipettes up to 2000ul, and all previous pipettes being converted to the XLS+ sealing system. Identical silicone lubricant sold by Rainin.