Learn More
Hach Company Pipet, Mohr, Glass, 10 mL
Supplier:  Hach Company 2093438
Encompass
Class B pipets calibrated “To Deliver”. Permanent brown, graduated scale fused into body. Color coded for volume. Tips hardened for durability. Designed from ASTM E1293, Class B Style 1 requirements.
Catalog No. 50-199-5346