Learn More
Chemglass Life Sciences Pasteur Pipet, Disposable Borosilicate Glass, 9 in, Pack of 200
SDP
Catalog No. NC2192653
Encompass_Preferred
Disposable borosilicate glass Pasteur pipets designed for one-time, non-volumetric liquid transfers in laboratory and clinical workflows. These pipets include a constriction one inch below the top to facilitate plugging and are suitable for rapid transfer applications in bacteriology, immunology, hematology, serology, blood bank, and clinical chemistry.
  • Disposable borosilicate glass construction.
  • 9 in length for extended reach.
  • Pack of 200 for bulk use.
  • Non-sterile, non-plugged design.
  • Constricted neck one inch below the top to facilitate plugging.
  • Suitable for rapid non-volumetric transfers in clinical and microbiology workflows.
This item is not returnable. View return policy
Catalog No. NC2192653 Supplier Chemglass Life Sciences Supplier No. CG87039
Disposable borosilicate glass Pasteur pipets designed for one-time, non-volumetric liquid transfers in laboratory and clinical workflows. These pipets include a constriction one inch below the top to facilitate plugging and are suitable for rapid transfer applications in bacteriology, immunology, hematology, serology, blood bank, and clinical chemistry.
  • Disposable borosilicate glass construction.
  • 9 in length for extended reach.
  • Pack of 200 for bulk use.
  • Non-sterile, non-plugged design.
  • Constricted neck one inch below the top to facilitate plugging.
  • Suitable for rapid non-volumetric transfers in clinical and microbiology workflows.