Learn More
Hach Company Pipet, Serological, Sterile, Glass, 10 mL, 25/pk
Supplier:  Hach Company 2093128
Encompass
Sterile, glass pipet with blue graduation scale. Calibrated “To Deliver”. Complies with ASTM E438, Class B requirements and designed from ASTM Specification E714.
Catalog No. 50-199-5339