Learn More
Hach Company Pipet, Serological, Sterile, Polystyrene with Plug, 1 mL, 1000/pk
Catalog No. 501995336 Shop All Hach Company Products
Encompass
Sterile, disposable plastic pipet with plug. Bulk packaged in 25/pack Color coded. Black ascending/descending graduations. Calibrated “To Deliver”. Accuracy ±2%.
Catalog No. 50-199-5336 Supplier Hach Company Supplier No. 2092855
Sterile, disposable plastic pipet with plug. Bulk packaged in 25/pack Color coded. Black ascending/descending graduations. Calibrated “To Deliver”. Accuracy ±2%.