Learn More
Hach Company Pipet, Serological, Sterile, Polystyrene, 1mL, Yellow, 1000/pk
Catalog No. 501995329 Shop All Hach Company Products
Encompass
Sterile, disposable plastic pipet. Individually wrapped. Color coded. Calibrated “To Deliver”. Complies with ASTM E934.
Catalog No. 50-199-5329 Supplier Hach Company Supplier No. 2092655
Sterile, disposable plastic pipet. Individually wrapped. Color coded. Calibrated “To Deliver”. Complies with ASTM E934.