Learn More
Hach Company Pipet, Serological, Sterile, Polystyrene, 5 mL, Blue, 200/pk
Catalog No. 501995331 Shop All Hach Company Products
Encompass
Sterile, disposable plastic pipet. Individually wrapped. Color coded. Calibrated “To Deliver”. Complies with ASTM E934.
Catalog No. 50-199-5331 Supplier Hach Company Supplier No. 2092667
Sterile, disposable plastic pipet. Individually wrapped. Color coded. Calibrated “To Deliver”. Complies with ASTM E934.