Learn More
Hach Company Pipet, Serological, Sterile, Polystyrene, 5 mL, Blue, 50/pk
Catalog No. 501995327 Shop All Hach Company Products
Encompass
Sterile, disposable plastic pipet. Individually wrapped. Color coded. Calibrated “To Deliver”. Complies with ASTM E934.
Catalog No. 50-199-5327 Supplier Hach Company Supplier No. 2092627
Sterile, disposable plastic pipet. Individually wrapped. Color coded. Calibrated “To Deliver”. Complies with ASTM E934.