Learn More
Wilmad Labglass PIPET SERO CL B 10ML ORG BD
Catalog No. 50802833
Encompass_Preferred
Serological Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 10mL. Subdivision: 0.1mL. Capacity Tolerance: 0.06mL. Color Band: Orange. Calibrated to deliver at 20degreeC. Pipet has permanent amber scale.
Catalog No. 50-802-833 Supplier Wilmad Labglass Supplier No. LG9335118
Serological Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 10mL. Subdivision: 0.1mL. Capacity Tolerance: 0.06mL. Color Band: Orange. Calibrated to deliver at 20degreeC. Pipet has permanent amber scale.