Learn More
Chemglass Life Sciences Pipet, Serological, 1mL, 280mm, Color Coded Yellow, Bulk Packed, Sterile
SDP
Catalog No. 501897599
Encompass
• IncludeS negative graduations for extra volume• Edged color-coded band for identification• Non-pyrogenic, non-cytotoxic, non-hemolytic, DNase- and RNase-free• Polyolefin fiber filter to reduce contaminationSerological pipettes can be used in both tissue culture and bacterial culture experiments. Made from high clarity polystyrene. Sterilized by E-beam.
Catalog No. 50-189-7599 Supplier Chemglass Life Sciences Supplier No. CGE3481001
• IncludeS negative graduations for extra volume• Edged color-coded band for identification• Non-pyrogenic, non-cytotoxic, non-hemolytic, DNase- and RNase-free• Polyolefin fiber filter to reduce contaminationSerological pipettes can be used in both tissue culture and bacterial culture experiments. Made from high clarity polystyrene. Sterilized by E-beam.