Learn More
Hach Company Pipet, Serological, Glass, 25 mL, 12/pk
Supplier:  Hach Company 206670
Encompass
Graduated glass pipets. Calibrated “To Deliver”. Designed to ASTM E1044 specifications.
Catalog No. 50-199-4672