Learn More
Chemglass Life Sciences Pipet, Serological, 5mL, 340mm, Color Coded Blue, Individually Wrapped, Sterile
SDP
Supplier:  Chemglass Life Sciences CGE3481005
Encompass
• Includes negative graduations for extra volume• Edged color-coded band for identification• Non-pyrogenic, non-cytotoxic, non-hemolytic, DNase- and RNase-free• Polyolefin fiber filter to reduce contaminationSerological pipettes can be used in both tissue culture and bacterial culture experiments. Made from high clarity polystyrene. Sterilized by E-beam.
Catalog No. 50-189-7603