Learn More
Hach Company Sterile Pipet, 10-11 mL, pk/25
Supplier:  Hach Company 209798
Encompass
Pipet, Serological, Sterile, 10-11 mL, 0.1 mL divisions, pk/25, Polystyrene, calibrated to deliver/blow-out. Corning.
Catalog No. 50-199-4679