Learn More
CELLTREAT Scientific Products Pipet Storage Rack
SDP
Catalog No. 50828753
Encompass_Preferred
CELLTREAT Scientific Products Pipet Storage Rack. Durable ABS plastic rack can stand on flat surfaces as well as be attached to the metal sides of lab cabinets and appliances. Strong magnets are included on both sides for increased versatility. Angled shelves provide secure yet easy access to pipets. Pipet sizes, colors, and types can be viewed through the side windows. Side bracket can hold pipet fillers, bulbs and pumps. Every case is printed with the lot number for quality traceability. Packaged 1/case.
Catalog No. 50-828-753 Supplier CELLTREAT Scientific Products Supplier No. 230295
CELLTREAT Scientific Products Pipet Storage Rack. Durable ABS plastic rack can stand on flat surfaces as well as be attached to the metal sides of lab cabinets and appliances. Strong magnets are included on both sides for increased versatility. Angled shelves provide secure yet easy access to pipets. Pipet sizes, colors, and types can be viewed through the side windows. Side bracket can hold pipet fillers, bulbs and pumps. Every case is printed with the lot number for quality traceability. Packaged 1/case.