Learn More
Labcon North America ZAP™ SLIK 10uL Extra Long Low Retention Aerosol Filter Pipet Tips, in Racks, Sterile
Catalog No. NC0864708
Encompass_Preferred
Tip, Pipet; Labcon; ZAP™ SLIK Low Retention Aerosol Filter; Extra length; Fits most popular 10uL pipettors; Ultra low fluid retention; Sterile; Patented robotically inserted endotoxin free filter; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Refillable autoclavable racks; 96 tips/rack; 10 racks/pack; 10 packs/case; 47.6mm H.; Made in USA
Catalog No. NC0864708 Supplier Labcon North America Supplier No. 11739650089
Tip, Pipet; Labcon; ZAP™ SLIK Low Retention Aerosol Filter; Extra length; Fits most popular 10uL pipettors; Ultra low fluid retention; Sterile; Patented robotically inserted endotoxin free filter; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Refillable autoclavable racks; 96 tips/rack; 10 racks/pack; 10 packs/case; 47.6mm H.; Made in USA