Learn More
Research Products International Corp UNIVERSAL PLUS PIPET TIPS, YELLOW, QUICKRACK, 20-200UL, NON-STERILE, 960/CASE
SDP
Catalog No. 50998489
Encompass
Our new and unique UniversalPlus Tips fit virtually all 20ul-200ul pipettors with a standard barrel. Laboratories can now standardize on one tip and eliminate the need for stocking many different types. The soft fit collar forms a secure and reliable seal and allows tips to be ejected with less effort. This feature reduces hand fatigue and improves efficiency with mult-channel pipettors. Tips have a fine point for precise delivery and graduation marks at 10ul, 50ul and 100ul. Maximum volume is 240ul. Available non-sterile.
Catalog No. 50-998-489 Supplier Research Products International Corp Supplier No. 131452
Our new and unique UniversalPlus Tips fit virtually all 20ul-200ul pipettors with a standard barrel. Laboratories can now standardize on one tip and eliminate the need for stocking many different types. The soft fit collar forms a secure and reliable seal and allows tips to be ejected with less effort. This feature reduces hand fatigue and improves efficiency with mult-channel pipettors. Tips have a fine point for precise delivery and graduation marks at 10ul, 50ul and 100ul. Maximum volume is 240ul. Available non-sterile.