Learn More
Bio Rad Laboratories PIPET TIPS FOR THE AUTODG SYST
Catalog No. NC0843491
Encompass
Package of 40, boxes of 96 each, pipet tips for use with the Automated Droplet Generator in the QX200 AutoDG Droplet Digital PCR System.
This item is not returnable. View return policy
Catalog No. NC0843491 Supplier Bio Rad Laboratories Supplier No. 1864121
Package of 40, boxes of 96 each, pipet tips for use with the Automated Droplet Generator in the QX200 AutoDG Droplet Digital PCR System.