Learn More
Research Products International Corp UniFit Pipet Tips, Sterile, Blue, 1000 µl, 10 Racks of 100, 1000 per Case
SDP
Catalog No. NC2608790
Encompass
Blue Pipet Tips fit all popular brands of pipettors including Pipetman, Eppendorf, Finnpipette, Socorex, MLA, Volac, Octapette and Titertek. Blue tips have a unique beveled end that minimizes fluid retention and improves accuracy. Available in a fully enclosed rack that protects tips from airborne contamination and allows repeated autoclaving up to 120°C Supplied sterile in rack of 100 tips; 10 racks; 1000/cs.
Catalog No. NC2608790 Supplier Research Products International Corp Supplier No. 141161
Blue Pipet Tips fit all popular brands of pipettors including Pipetman, Eppendorf, Finnpipette, Socorex, MLA, Volac, Octapette and Titertek. Blue tips have a unique beveled end that minimizes fluid retention and improves accuracy. Available in a fully enclosed rack that protects tips from airborne contamination and allows repeated autoclaving up to 120°C Supplied sterile in rack of 100 tips; 10 racks; 1000/cs.