Learn More
Andwin Scientific PIPET TRANSFER 23ML 12 PK/100
SDP
Catalog No. NC9075706
Encompass_Preferred
The disposable 23 mL transfer pipet with extra long tip is made from see-through low density polyethylene that is long-lasting as it will not shatter or break. This product is classified as nontoxic and inert to most acids. It is also compatible to use for liquid oxygen. This transfer pipet features a uniform drop size and a low-affinity surface that minimizes the loss of cells and important proteins as a result of binding.
Catalog No. NC9075706 Supplier Andwin Scientific Supplier No. 16001196
The disposable 23 mL transfer pipet with extra long tip is made from see-through low density polyethylene that is long-lasting as it will not shatter or break. This product is classified as nontoxic and inert to most acids. It is also compatible to use for liquid oxygen. This transfer pipet features a uniform drop size and a low-affinity surface that minimizes the loss of cells and important proteins as a result of binding.