Learn More
Thomas Scientific Transfer Pipet, 3mL, short bulb, grad. to 1mL
SDP
Catalog No. 501460829
Encompass_Preferred
Produced from a custom grade of low-density polyethylene (LDPE), these unbreakable one-piece pipets are used to draw and dispense liquids in controlled, reproducible drop sizes. Transfer Pipet, 3mL short bulb, bulk grad. to 1mL 135mm 250/pk 5000/cs (20pk)
Catalog No. 50-146-0829 Supplier Thomas Scientific Supplier No. 1158Q82
Produced from a custom grade of low-density polyethylene (LDPE), these unbreakable one-piece pipets are used to draw and dispense liquids in controlled, reproducible drop sizes. Transfer Pipet, 3mL short bulb, bulk grad. to 1mL 135mm 250/pk 5000/cs (20pk)