Learn More
Thomas Scientific Dropette Transfer Pipet, 3mL, 13.8cm Length, 2.1mL Bulb Draw, Non-Sterile, Graduated (Loose/Pack)
SDP
Catalog No. 501460816
Encompass
Made of low density polyethylene These all-in-one pipets eliminate the hazard of broken glass and exposure to infectious materials. Put an end to matching rubber bulbs with glass pipets. Molded from see-through low density polyethylene. Inert to biological fluids and most acids. Can be sealed and refrigerated. They work well whenever there is a need for quick, safe transfer of fluids. Temperature resistant down to -196°C. Can be gaz (EtO) sterilized. Choose between 12 very popular models, available in several sizes, tip designs and lengths, in sterile or non sterile packaging. Seven models provide graduations. Will not shatter Can be used in liquid nitrogen Non toxic and inert No bulb to insert or remove Uniform drop size Packaging: All non-sterile pipets are in boxes of 500 and cases of 5000. Exception: P200-82 are in boxes of 400 and cases of 4000. Most sterile pipets are in boxes of 400 and cases of 4000.
Catalog No. 50-146-0816 Supplier Thomas Scientific Supplier No. 1219D73
Made of low density polyethylene These all-in-one pipets eliminate the hazard of broken glass and exposure to infectious materials. Put an end to matching rubber bulbs with glass pipets. Molded from see-through low density polyethylene. Inert to biological fluids and most acids. Can be sealed and refrigerated. They work well whenever there is a need for quick, safe transfer of fluids. Temperature resistant down to -196°C. Can be gaz (EtO) sterilized. Choose between 12 very popular models, available in several sizes, tip designs and lengths, in sterile or non sterile packaging. Seven models provide graduations. Will not shatter Can be used in liquid nitrogen Non toxic and inert No bulb to insert or remove Uniform drop size Packaging: All non-sterile pipets are in boxes of 500 and cases of 5000. Exception: P200-82 are in boxes of 400 and cases of 4000. Most sterile pipets are in boxes of 400 and cases of 4000.