Learn More
Thomas Scientific Transfer Pipet, 7mL, lg. bulb, grad. to 3mL
SDP
Catalog No. 501460828
Encompass_Preferred
Produced from a custom grade of low-density polyethylene (LDPE), these unbreakable one-piece pipets are used to draw and dispense liquids in controlled, reproducible drop sizes. Transfer Pipet, 7mL lg. bulb, bulk grad. to 3mL 160mm 250/pk 5000/cs (20pk)
Catalog No. 50-146-0828 Supplier Thomas Scientific Supplier No. 1158Q83
Produced from a custom grade of low-density polyethylene (LDPE), these unbreakable one-piece pipets are used to draw and dispense liquids in controlled, reproducible drop sizes. Transfer Pipet, 7mL lg. bulb, bulk grad. to 3mL 160mm 250/pk 5000/cs (20pk)