Learn More
Wilmad Labglass PIPET VOL CLASS A 12.5ML
Catalog No. 50807837
Encompass
Volumetric Pipet, Class 'A', To Deliver, Color-Coded. Capacity: 12.5mL. Capacity Tolerance: 0.03mL. Color Band: Orange. Manufactured to meet the requirements for accuracy,appearance,glass quality and calibration line accuracy of ASTM E969.
Catalog No. 50-807-837 Supplier Wilmad Labglass Supplier No. LG9350119
Volumetric Pipet, Class 'A', To Deliver, Color-Coded. Capacity: 12.5mL. Capacity Tolerance: 0.03mL. Color Band: Orange. Manufactured to meet the requirements for accuracy,appearance,glass quality and calibration line accuracy of ASTM E969.