Learn More
Wilmad Labglass PIPET VOL CLASS A 15ML
Supplier:  Wilmad Labglass LG9350120
Encompass_Preferred
Volumetric Pipet, Class 'A', To Deliver, Color-Coded. Capacity: 15mL. Capacity Tolerance: 0.03mL. Color Band: Green. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.
Catalog No. 50-807-526