Learn More
Wilmad Labglass PIPET VOL CLASS A 1ML
Catalog No. 50807884
Encompass_Preferred
Volumetric Pipet, Class 'A', To Deliver, Color-Coded. Capacity: 1mL. Capacity Tolerance: 0.006mL. Color Band: Blue. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.
Catalog No. 50-807-884 Supplier Wilmad Labglass Supplier No. LG9350101
Volumetric Pipet, Class 'A', To Deliver, Color-Coded. Capacity: 1mL. Capacity Tolerance: 0.006mL. Color Band: Blue. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.