Learn More
Wilmad Labglass PIPET VOL CLASS A 6ML
Supplier:  Wilmad Labglass LG9350110
Encompass_Preferred
Volumetric Pipet, Class 'A', To Deliver, Color-Coded. Capacity: 6mL. Capacity Tolerance: 0.01mL. Color Band: 2 Orange. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.
Catalog No. 50-807-892