Learn More
Hach Company Pipet, Volumetric, Class A, Certified, 2 mL, 6/pk
Supplier:  Hach Company 2636166
Encompass
Glass Class A pipets serialized and calibrated “To Deliver”. Each comes with Certificate of Identification and Capacity. Marked with an “A” to designate compliance to ASTM E969.
Catalog No. 50-199-6407