Learn More
Wilmad Labglass PIPET VOL CLASS B 100ML
Supplier:  Wilmad Labglass LG9355136
Encompass_Preferred
Volumetric Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 100mL. Capacity Tolerance: 0.16mL. Color Band: Yellow. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.
Catalog No. 50-806-081